Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes™ Secondary Antibody, Alexa Fluor™ 488 - FAQs
Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes™ Secondary Antibody, Alexa Fluor™ 488 - FAQs
View additional product information for Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes™ Secondary Antibody, Alexa Fluor™ 488 - FAQs (R37116)
1 product FAQs found
Can I use Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes Secondary Antibody (Cat. No. R37116) on tissue sections (both paraffin-embedded or cryosections)?
Yes, Goat anti-Rabbit IgG (H+L) Cross-Adsorbed ReadyProbes Secondary Antibody (Cat. No. R37116) can be used on tissue sections (both paraffin-embedded or cryosections) but you may need a longer incubation time than indicated in the quick protocol. Two hours incubation at room temperature is the typical process, but we recommend optimizing incubation times with control samples.