ReadyProbes™ Cell Viability Imaging Kit, Blue/Red - FAQs
ReadyProbes™ Cell Viability Imaging Kit, Blue/Red - FAQs
View additional product information for ReadyProbes™ Cell Viability Imaging Kit, Blue/Red - FAQs (R37610)
1 product FAQs found
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.