ReadyProbes™ Mouse-on-Mouse IgG Blocking Solution (30X) - FAQs
ReadyProbes™ Mouse-on-Mouse IgG Blocking Solution (30X) - FAQs
View additional product information for ReadyProbes™ Mouse-on-Mouse IgG Blocking Solution (30X) - FAQs (R37621)
4 product FAQs found
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Can I use ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) on rat tissues?
Due to some cross-reactivity of anti-mouse antibodies to rat antibodies, using ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) on rat tissues may offer some level of blocking, but it may not be sufficient. For rat tissues, ideally you should use a rat-on-rat blocking.
Can I combine the standard blocking step (using serum or BSA) with the ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) to save time?
No. These two blocking steps cannot be combined. Doing so would result in inadequate blocking for both non-specific entities and mouse antibodies.
Do I need to use ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) on cultured mouse cells?
Assuming that the cultured mouse cells are not expressing mouse antibodies, ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) is not needed for cultured mouse cells.