ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X)
Invitrogen™
ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X)
ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activity in formalin-fixed paraffin-embedded tissueRead more
Have Questions?
Catalog NumberQuantity
R37629100 mL
Catalog number R37629
Price (USD)
181.22
Special offer
Online exclusive
Ends: 31-Dec-2026
221.00
Save 39.78
Each
Quantity:
100 mL
Price (USD)
181.22
Special offer
Online exclusive
Ends: 31-Dec-2026
221.00
Save 39.78
Each
ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activity in formalin-fixed paraffin-embedded tissue sections, frozen tissue sections, and cell preparations. It is provided in a ready-to-use format—just add a few drops and incubate for 10–15 minutes. Endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activities become problematic when alkaline phosphatase (AP)- or horseradish peroxidase (HRP)- based detection systems are used. ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits these endogenous activities, for reduced non-specific signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures. This blocker is used before any standard, non-specific blocking solutions (e.g., diluted serum).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product TypeBlocking Solution
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product LineReadyProbes
Quantity100 mL
Unit SizeEach
Contents & Storage
Provided as 100-mL solution at ready-to-use (1X) concentration in a white dropper bottle.
Store at 2–8°C.
When stored as directed, product can be used for 6 months after receipt.
Frequently asked questions (FAQs)
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can ReadyProbes Endogenous HRP and AP Blocking Solution (1X) be used on cell or tissue lysates?
We do not recommend using ReadyProbes Endogenous HRP and AP Blocking Solution (1X) on cell or tissue lysates. The ReadyProbes Endogenous HRP and AP Blocking Solution (1X) must be removed prior to the application of any HRP- or AP-conjugated antibody/streptavidin. If analyzing the lysates directly, the ReadyProbes Endogenous HRP and AP Blocking Solution (1X) cannot be removed from the lysate.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can ReadyProbes Endogenous HRP and AP Blocking Solution (1X) be used on live cells?
ReadyProbes Endogenous HRP and AP Blocking Solution (1X) cannot be used on live cells.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How does ReadyProbes Endogenous HRP and AP Blocking Solution (1X) work?
ReadyProbes Endogenous HRP and AP Blocking Solution (1X) works by irreversibly inactivating the enzymes.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.