ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X) - FAQs
ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X) - FAQs
View additional product information for ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X) - FAQs (R37629)
4 product FAQs found
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Can ReadyProbes Endogenous HRP and AP Blocking Solution (1X) be used on cell or tissue lysates?
We do not recommend using ReadyProbes Endogenous HRP and AP Blocking Solution (1X) on cell or tissue lysates. The ReadyProbes Endogenous HRP and AP Blocking Solution (1X) must be removed prior to the application of any HRP- or AP-conjugated antibody/streptavidin. If analyzing the lysates directly, the ReadyProbes Endogenous HRP and AP Blocking Solution (1X) cannot be removed from the lysate.
Can ReadyProbes Endogenous HRP and AP Blocking Solution (1X) be used on live cells?
ReadyProbes Endogenous HRP and AP Blocking Solution (1X) cannot be used on live cells.
How does ReadyProbes Endogenous HRP and AP Blocking Solution (1X) work?
ReadyProbes Endogenous HRP and AP Blocking Solution (1X) works by irreversibly inactivating the enzymes.