ReadyProbes™ Hydrophobic Barrier Pap Pen - FAQs
ReadyProbes™ Hydrophobic Barrier Pap Pen - FAQs
View additional product information for ReadyProbes™ Hydrophobic Barrier Pap Pen - FAQs (R3777)
1 product FAQs found
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.