COPAN WASP™DT Instrument Accessories and Software
COPAN WASP™DT Instrument Accessories and Software
Build a more efficient microbiology laboratory and automate all aspects of specimen processing with COPAN WASP™DT Instrument Accessories and Software.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
R5086031Plate Side Label6000/Roll
R5086030Plate Bottom Label4000/Roll
R5086032Printer Ink Ribbon1 Roll
R5086035Sterile Empty Tubes500/Box
R5086036Drip Trays24/Box
R5086050Electric Drip Trays128/Box
R5086039Specimen Rejection Tray Foam Liner40/Box
R5086040Loop Cleaning Solution50/Box
R5086042Gram Slides1500/Box
R5086043Gram SlidePrep Ink Cartridge1 Ea.
R5086044Biquetra 1μL Loop2/Box
R5086045Biquetra 10μL Loop2/Box
R5086046Biquetra 30μL Loop2/Box
R5086047Twin Biquetra 1μL Loop1 Ea.
R5086048Gram SlidePrep Drip Trays200/Box
R5086049Docking Station Pads36/Box
Catalog number R5086031
Price (USD)
-
Product Type:
Plate Side Label
Quantity:
6000/Roll
Build a more efficient microbiology laboratory and automate all aspects of specimen processing with COPAN WASP™DT Instrument Accessories and Software. Structure the modular system according to the unique needs of your laboratory, and choose from a variety of instrument accessories, from enrichment broth inoculation to an automatic loop and tool changing station, to drive productivity from start to finish as well as maximize current and future investments.
The WASP™DT System works seamlessly with Thermo Scientific™ Remel™ prepared media products which are optimized for automation.
Addressing all aspects of microbiology specimen processing with standardized automation for consistent quality, the WASP™DT System's modular platform ensures optimized lab productivity.
  • Up to 378 plate capacity increases walk-away time—for high-throughput and faster returns on investment, use Thermo Scientific Remel bi-plates
  • Multi-media carousel processes various culture setups using up to nine different types of media
  • Dynamic pick and place robotics mimic workflow routinely performed by lab technicians and prevents cross contamination
  • Continuous and random loading eliminates the need for batching
  • Specimen verification system ensures quality
  • Smart Scan Technology reduces errors and streamlines traceability
  • Proprietary gel-preservation ensures faster recovery with Thermo Scientific™ Culti-Loops™ Quality Control Organisms
Get the most out of your automated specimen processing system with Thermo Scientific Remel prepared media products. Manufactured in state-of-the-art facilities and supported by a responsive team of microbiology experts, Remel prepared media is the preferred choice of laboratories around the world. With more than 1,700 formats and formulations designed to deliver unmatched recovery and reproducibility, you can rest assured knowing that your automated specimen processing is working with products of the highest quality and consistency.
COPAN WASP™DT instrumentation and accessories are available through Thermo Fisher Scientific in the U.S.A. only. Please contact your local sales representative for more information.
WASP™DT is a registered trademark of COPAN S.P.A. All other trademarks are property of Thermo Fisher Scientific and its subsidiaries.
Specifications
Product LineWASP™DT
Quantity6000/Roll
Product TypePlate Side Label
Unit SizeEach