Product Image
Invitrogen™
Polypropylene Columns (Empty)
These empty, 10-mL polypropylene columns are for use with the ProBond™ Purification System and hold ProBond™ resin.Read more
Have Questions?
Catalog NumberQuantity
R6405050 Columns
Catalog number R64050
Price (USD)
409.00
Each
Quantity:
50 Columns
Request bulk or custom format
Price (USD)
409.00
Each
These empty, 10-mL polypropylene columns are for use with the ProBond™ Purification System and hold ProBond™ resin.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatEmpty Column
Product TypePolypropylene Column
Quantity50 Columns
Unit SizeEach
Contents & Storage
Store at room temperature.
Citations & References (2)
Citations & References
Abstract
Total synthesis of cyclic ADP-carbocyclic-ribose, a stable mimic of Ca2+-mobilizing second messenger cyclic ADP-ribose.
Authors: Shuto S; Fukuoka M; Manikowsky A; Ueno Y; Nakano T; Kuroda R; Kuroda H; Matsuda A;
Journal:J Am Chem Soc
PubMed ID:11535079
'The synthesis of cyclic ADP-carbocyclic-ribose (cADPcR, 4) designed as a stable mimic of cyclic ADP-ribose (cADPR, 1), a Ca2+-mobilizing second messenger, was achieved using as the key step a condensation reaction with the phenylthiophosphate-type substrate 14 to form an intramolecular pyrophosphate linkage. The N-1-carbocyclic-ribosyladenosine derivative 16 was prepared via the ... More
ProNGF induces p75-mediated death of oligodendrocytes following spinal cord injury.
Authors:Beattie MS, Harrington AW, Lee R, Kim JY, Boyce SL, Longo FM, Bresnahan JC, Hempstead BL, Yoon SO,
Journal:Neuron
PubMed ID:12408842
The neurotrophin receptor p75 is induced by various injuries to the nervous system, but its role after injury has remained unclear. Here, we report that p75 is required for the death of oligodendrocytes following spinal cord injury, and its action is mediated mainly by proNGF. Oligodendrocytes undergoing apoptosis expressed p75, ... More