Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56790. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ARHGAP18 is one member of the human RhoGAP family with approximately 80 RhoGAP proteins known to be encoded in the human genome. Rho proteins belong to the Ras superfamily that is composed of over 50 members divided into six families, including Ras, Sar, Rho, Ran, Rab and Arf. Rho GTPases are important regulators of the actin cytoskeleton and consequently influence the shape and migration of cells. ARHGAP18 is linked to Ras, and thus, to EGFR-mediated proliferation, migration and differentiation. ARHGAP18 is precisely contained within chromosome 6q22-24, which has been shown to be linked to schizophrenia, suggesting that ARHGEP18 may play a role in this condition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: rho-type GTPase-activating protein 18; rhoGAP
Gene Aliases: bA307O14.2; MacGAP; SENEX
UniProt ID: (Human) Q8N392
Entrez Gene ID: (Human) 93663
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support