Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61434. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NAALADL1 has NAALADase-like activity which is currenty unknown in function. NAALADL1 has no NAAG hydrolyzing activity, exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 100 kDa ileum brush border membrane protein; human ileal aminopeptidase; I100; ileal dipeptidylpeptidase; ileal peptidase I100; N-acetylated-alpha-linked acidic dipeptidase-like protein; NAALADase L; NAALADase-like 1; peptidase homolog
Gene Aliases: HILAP; I100; NAALADASEL
UniProt ID: (Human) Q9UQQ1
Entrez Gene ID: (Human) 10004
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support