Mammary Epithelial Growth Supplement (MEGS) - FAQs
Mammary Epithelial Growth Supplement (MEGS) - FAQs
View additional product information for Mammary Epithelial Growth Supplement (MEGS) - FAQs (S0155)
1 product FAQs found
Is Mammary Epithelial Growth Supplement (MEGS) suitable for MCF10A culture?
Yes, Mammary Epithelial Growth Supplement (MEGS) is suitable for MCF10A culture. You can also include 5% horse serum if desired.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.