The SiteClick™ sDIBO Alkyne Kits are optimized to be used in conjunction with our SiteClick™ Duet or Quartet Antibody Labeling Kits.
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
Invitrogen™

SiteClick™ sDIBO Alkyne Kits for Antibody Labeling

Easily attach Alexa Fluor™ pHrodo™ and Biotin sDIBO Alkyne tags to azido-modified antibodies using SiteClick™ chemistry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or DyeLabeling Scale
S10915pHrodo Deep Red1 mg
C20027Alexa Fluor 488100-250 μg
C20029Alexa Fluor 647100-250 μg
C20031Primary Amine100-250 μg
C20034pHrodo Red100-250 μg
S10902Biotin1 mg
S10903pHrodo Red1 mg
S10904Alexa Fluor 4881 mg
S10906Alexa Fluor 6471 mg
S10907Biotin5 mg
S10908pHrodo Red5 mg
S10909Alexa Fluor 4885 mg
S10911Alexa Fluor 6475 mg
S10914pHrodo Deep Red100-250 μg
C20028Alexa Fluor 555100-250 μg
C20030Biotin100-250 μg
Catalog number S10915
Price (USD)
700.00
Each
Label or Dye:
pHrodo Deep Red
Labeling Scale:
1 mg
Request bulk or custom format
Price (USD)
700.00
Each
Generate site-specific labeled antibody conjugates with SiteClick sDIBO Alkynes for Antibody Labeling. Our SiteClick sDIBO Antibody Labeling kits are optimized to be used in conjunction with the SiteClick Azido Modification kits (S20026, S10900, or S10901 sold separately) and contain sufficient reagents to perform a conjugation reaction of a SiteClick sDIBO Alkyne label using 100 μg, 1 mg, or 5 mg of azido-modified IgG antibody. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor dye, pHrodo dye, or biotin to an azido modified antibody. The label, when attached to the antibody, is located far away from its antigen binding domain.

Create your own covalently labeled antibody conjugates using the SiteClick sDIBO Alkyne labels, which are intended for use with azido-modified antibodies. SiteClick sDIBO Alkyne Antibody Labeling Kits include the sDIBO Alkyne and the antibody concentrator, enabling copper-free covalent attachment of Alexa Fluor and pHrodo dyes, biotin, or molecules of your choice. When used with the SiteClick Antibody Azido Modification Kit (available separately as Cat. No. S20026, S10900, S10901), labeling occurs on the heavy chain N-linked glycans, far away from the antigen-binding domain, enabling excellent reproducibility between separate labeling reactions and multiple antibody species and subtypes. The SiteClick sDIBO alkyne kits for antibody labeling are optimized for conjugation reactions, covalently linking 100 μg, 1 mg, or 5 mg of azide-modified IgG antibody.

The SiteClick Antibody Azido Modification Kit (Cat. No. S20026, S10900, S10901) is recommended for the attachment of an azide moiety to the heavy chains of an unlabeled IgG antibody. Other engineered antibodies containing azido moieties are also suitable.

When used in conjunction with the SiteClick Antibody Azido Modification Kit, the SiteClick sDIBO alkyne kits for antibody labeling give you the option of trying different fluorescent labels for your antibody, attaching other molecules via streptavidin, or attaching your own molecule via amine-reactive moieties, depending on your assay. Our sDIBO alkynes are improved versions of DIBO cyclooctynes, yielding conjugates that are less 'sticky' and emit lower signal background in biological samples.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAzide
Excitation/Emission640/655 nm
Filter TypeCy5
Flow Cytometer Laser LinesRed
Kit ContentsContains 1 vial of SiteClick pHrodo Deep Red sDIBO Alkyne for Antibody Labeling in 50 μL DMSO, 1 dye removal column, 1 wash vial, 1 collection vial, and PBS exchange buffer
Labeling MethodClick Chemistry
Labeling Scale1 mg
Labeling TargetAzido modified antibodies
Label or DyepHrodo Deep Red
QuantityFor up to 1 mg of Ab
Reactive MoietyAlkyne
Shipping ConditionWet Ice
ColorDeep Red
Product LinepHrodo, SiteClick
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

May I store antibodies or other proteins labeled with SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915) in solutions containing sodium azide?

We do not recommend storing antibodies or proteins labeled with SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915) in solutions containing sodium azide, as pHrodo Deep Red is not stable in the presence of sodium azide. Consider using an alternative, non-azide-based anti-microbial.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How can I perform a DOL determination for SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915)?

To determine the Degree of Labeling (DOL) for the SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915), refer to page 13 of the SiteClick Antibody Azido Modification Kit User Guide.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

May I use the SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915), to label any azide-modified protein besides antibodies?

Yes. You can use the SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915), to label any azide-modified protein as long as the azide moiety is accessible. You may perform a copper-free click reaction to label the protein with the sDIBO alkynes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.