Secure-Seal™ Spacer, one well, 13 mm diameter, 0.12 mm deep - FAQs
Secure-Seal™ Spacer, one well, 13 mm diameter, 0.12 mm deep - FAQs
View additional product information for Secure-Seal™ Spacer, one well, 13 mm diameter, 0.12 mm deep - FAQs (S24735)
1 product FAQs found
I would like to know what quantity corresponds to 1 set of Secure-Seal Spacer, one well, 13 mm diameter, 0.12 mm deep (Cat. No. S24735)?
It contains 100 spacers.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center