For Streptavidin, Allophycocyanin Conjugate, premium grade (Cat. No. S32362), how many molecules of APC are conjugated to each streptavidin molecule?
Streptavidin APC, premium grade has a degree of labeling of one APC molecule to one streptavidin molecule.