SlowFade™ Gold Antifade Mountant with DAPI, 2 mL - FAQs
SlowFade™ Gold Antifade Mountant with DAPI, 2 mL - FAQs
View additional product information for SlowFade™ Gold Antifade Mountant with DAPI - FAQs (S36938, S36939, S36942)
2 product FAQs found
Does Slowfade Antifade Mountant require using a nail polish for sealing?
No. Samples mounted with SlowFade mountants need not be sealed; they are intended for immediate viewing. The coverslip may be anchored (to prevent movement while viewing) by applying molten paraffin to three or four spots around the edge of the coverslip.
DAPI and Hoechst dyes are quite similar to each other. Why would I choose one over the other?
DAPI is a very common blue-fluorescent dye for nuclear counterstaining and gives very bright labeling on nuclei in fixed and permeabilized cells and tissues. However, it is considered to be a semi-permeant to impermeant stain and provides inconsistent staining of live cells. Hoechst 33342 dye is cell-permeant and stains with the same binding mechanism and fluorescent color; it is preferred for live-cell imaging and is just as good as DAPI for fixed cell labeling.