Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Donkey anti-Rabbit IgG Secondary Antibodies

          Identifyn

          F(ab')2-Donkey anti-Rabbit IgG, (H+L) SRM Identifyn™ Secondary Antibody, Alexa Fluor™ 555

          View all (73) Donkey anti-Rabbit IgG antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Datasheet
          Certificates
          Questions & Answers
          Datasheet
          Certificates
          Questions & Answers

          Cite F(ab')2-Donkey anti-Rabbit IgG, (H+L) SRM Identifyn™ Secondary Antibody, Alexa Fluor™ 555

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rabbit IgG, (H+L) SRM Secondary Antibody (SA000043) in ICC/IF

          IdentifynTM Detailed protocols can be found below under Documents & Downloads. Confocal Microscopy: HeLa Cells were grown on acid-etched 1.5H cover glass to ~50% confluency, followed by -20°C methanol fixation and blocked with Thermo Fisher, Fish Serum Blocker (Product # 37527X3). The first primary antibody, Mitochondria Rabbit Recombinant Antibody (MTC02,2860R), was incubated overnight at 4°C at a dilution... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG, (H+L) SRM Secondary Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          SA000043

          Applications
          Tested Dilution

          Western Blot (WB)

          1:2,000

          Immunocytochemistry (ICC/IF)

          1:250-1:1,000
          Product Specifications

          Species Reactivity

          Rabbit

          Host/Isotype

          Donkey / IgG

          Class

          Polyclonal

          Type

          Secondary Antibody

          Immunogen

          Proprietary Immunogen.

          Conjugate

          Alexa Fluor™ 555 Alexa Fluor™ 555 Alexa Fluor™ 555
          • Alexa Fluor 405 
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 750 
          • Alexa Fluor 790 
          • Request custom conjugation

          Excitation/Emission Max

          553/568 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 10% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target

          IgG

          Antibody Form

          F(ab')2 Fragment

          Product Specific Information

          Identifyn™ Primary and Secondary antibodies are extensively validated in Immunofluorescence assays by traditional as well as high resolution acquisition like Widefield SIM, Confocal, Airyscan and STORM Microscopy.

          Recommended dilution for traditional microscopy:

          Widefield - 1:1,000

          Widefield SIM - 1:1,000

          Confocal - 1:1,000

          Recommended dilution for Super Resolution microscopy:

          SIM (Structured Illumination Microscopy) - 1:1,000

          Single Molecule STORM (Stochastic Optical Reconstruction Microscopy) - 1:250

          Detailed protocols can be found below under Methods & Protocols.

          Target Information

          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody SA000043 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline