Streptavidin, (FITC) - FAQs
Streptavidin, (FITC) - FAQs
View additional product information for Streptavidin, (FITC) - FAQs (SA1001)
1 product FAQs found
Where can I find the antibody concentration for Streptavidin, (FITC) (Cat. No. SA1001)?
The antibody concentration for Streptavidin, (FITC) can be found in the Certificate of Analysis for your specific lot number.