SureClick™ Antibody Labeling Kit
Invitrogen™
SureClick™ Antibody Labeling Kit
Invitrogen SureClick Antibody Labeling Kits enable fast, precise, and reproducible conjugation of human or rabbit IgG antibodies using trusted click chemistry. Designed for spatial biology researchers and biotherapeutic developers, SureClick kits attach an average of 2 payloads per antibody at the Fc region.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SC0013 x 100 μg
SC0023 x 1 mg
SC0035 mg
Catalog number SC001
Price (USD)
600.00
Each
Quantity:
3 x 100 μg
Price (USD)
600.00
Each
Covalent, Fc-targeted antibody conjugation in just two hours
Invitrogen SureClick Antibody Labeling Kits enable fast, precise, and reproducible conjugation of human or rabbit IgG antibodies using trusted click chemistry. Designed for spatial biology researchers and biotherapeutic developers, SureClick kits attach an average of 2 payloads per antibody at the Fc region, preserving antigen binding and Fc receptor (FcR) affinity.
SureClick is an exceptional solution for attaching oligonucleotides to antibodies, making it ideal for spatial biology workflows, immuno-PCR, and next-generation antibody-based detection platforms that require precise and reproducible antibody–oligo conjugation.
Achieve consistent, covalent antibody labeling with a simplified workflow amenable to automation.
What is SureClick antibody labeling?
SureClick is a site-specific antibody conjugation technology that covalently attaches payloads—including fluorophores, functional tags, and oligonucleotides—to the Fc region of IgG antibodies using click chemistry. Unlike conventional random labeling approaches, SureClick:
  • Targets the Fc domain
  • Preserves Fab antigen-binding regions
  • Maintains Fc glycosylation integrity
  • Enables reproducible degree of labeling (DOL)
  • Supports consistent antibody–oligonucleotide conjugation
Key features and benefits:
Precise, reproducible labeling
  • Site-specific Fc conjugation
  • Consistent attachment of ~2 payloads per antibody
  • Reduced heterogeneity compared to conventional methods
Rapid two-hour workflow
  • ~10 minutes hands-on time
  • 90-minute protocol
  • Simple protocol using a single Fc-targeting component
Covalent and stable conjugation
  • Trusted click chemistry
  • Robust linkage for downstream assays, spatial biology, and antibody–oligo workflows
Preserves antibody function
  • Confidence that antigen binding is unchanged
  • Maintains Fc receptor interactions
Automation-ready
  • Spin-column purification
  • Minimal components
  • Suitable for high-throughput workflows
How SureClick antibody labeling works
  1. Add SureClick Fc-targeting reagent to your IgG antibody
  2. Purify using included Zeba™ spin columns
  3. Add click chemistry payload of choice (e.g., fluorophores, oligonucleotides, functional tags)
The result is a site-specific, covalently labeled antibody with consistent performance, including highly controlled antibody–oligonucleotide conjugates.
Applications
  • Spatial biology
  • Antibody–oligonucleotide conjugation
  • Multiplex biomarker detection
  • Antibody-drug conjugate (ADC) development
  • Biotherapeutic characterization
  • Flow cytometry
  • Internalization studies
  • Biomarker validation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Antibody Production and Labeling
For Use With (Equipment)Benchtop Centrifuge
FormatLabeling Kit
Incubation Time70 to 90 min.
Labeling MethodClick Chemistry
Labeling ScaleApprox. 2°
Product LineSureClick™
Product TypeAntibody Labeling Kit
Quantity3 x 100 μg
Reactive MoietyAzide
Shipping ConditionWet Ice
SpeciesHuman or Rabbit
Sufficient ForLabeling 3 Antibodies
Chemical ReactivityAzide
Labeling TargetAntibodies
Label or DyeAzide
SolubilityAqueous Buffer
SpacerMid-length
Unit SizeEach
Contents & Storage
SureClick™ Fc Targeting Reagent
2°C to 8°C
Frequently asked questions (FAQs)
What antibody types are compatible?
SureClick labeling is designed for human and rabbit IgG antibodies.
How long does SureClick labeling take?
The full protocol takes approximately 90 minutes with about 10 minutes of hands-on time.
Does SureClick labeling affect antigen binding?
No. The SureClick reaction targets the Fc region, preserving the Fab antigen-binding domain and maintaining Fc receptor interactions..
Is SureClick labeling suitable for attaching oligonucleotides to antibodies?
Yes. SureClick labeling is an exceptional method for antibody–oligonucleotide conjugation. Its Fc-specific, site-controlled labeling enables reproducible and precise attachment of oligonucleotides while preserving antigen binding and antibody functionality.
How many labels does the SureClick reagent attach per antibody?
The SureClick reagent consistently attaches approximately two payloads per antibody.