Search
| 製品番号(カタログ番号) | モデル |
|---|---|
| SENTINEL4000D | Sentinel 4000 Drop Through |
| SENTINEL4000P | Sentinel 4000 Pipeline |
Two models have been designed to address the needs of different applications. The drop-through system is well-suited for use with granular and powdered food products, cosmetics, and chemicals, while the pipeline system is engineered for slurries, emulsions, and other viscous or viscoelastic products.
The Sentinel 4000 systems can identify contaminants that are up to 50% smaller in size than the Thermo Scientific™ APEX Metal Detector. This helps manufacturers increase customer safety, improve product quality, and drive production efficiency.
Thermo Scientific Sentinel 4000 Systems come with Multiscan frequency technology that enables optimized performance in any application, with up to five user-selectable frequencies that allow for improved sensitivity and detection of smaller contaminants. This technology generates a higher probability of metal contaminant detection, leading to increased food safety and improved brand protection.
Notable Sentinel 4000 Drop Through System features
Notable Sentinel 4000 Pipeline System features