iBind PLUS Solution Kit
Invitrogen™

iBind PLUS Solution Kit

The iBind PLUS Solution Kit increases signal sensitivity, requires less antibody per run, and simplifies western blotting immunoprocessing when used with the iBind Western System. The iBind PLUS Solution Kit comes with two ready to use solutions, iBind PLUS Solution and iBind PLUS Antibody Diluent.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SLF10301 Kit
SLF1030X44 Kits
Catalog number SLF1030
Price (USD)
118.00
Each
Quantity:
1 Kit
Request bulk or custom format
Price (USD)
118.00
Each

The iBind PLUS Solution Kit increases signal sensitivity, requires less antibody per run, and simplifies western blotting immunoprocessing when used with the iBind Western System. The iBind PLUS Solution Kit comes with two ready to use solutions, iBind PLUS Solution and iBind PLUS Antibody Diluent.

  • The iBind PLUS Solution comes ready to use with no preparation required. It is used to prewet the iBind card prior to the run and acts as the block and wash buffer for the membrane processing, helping wash away unbound primary and secondary antibodies.
  • The iBind PLUS Antibody Diluent is used to dilute the primary and secondary antibodies. In most experiments, the primary antibody can be diluted to the same recommended concentration as used for traditional blotting, however only 2 mL of prepared primary antibody is required with iBind, saving up to 80% of primary antibody consumption. The iBind PLUS Antibody Diluent is specially formulated with bulking agents and stabilizing molecules to improve antibody binding with the target antigen to increase signal intensity.

During a run on the iBind, each solution is released from the iBind device wells through sequential lateral flow (SLF) and are wicked over the iBind card towards the stack region, incubating the membrane with each reagent. The glass fiber matrix of the iBind card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

The iBind PLUS Solution Kit, together with the iBind Western Device (Cat. No. SLF1000) and iBind Cards (Cat. No. SLF1010), bring automation and simplicity to western blotting with efficient and hands-free immunoprocessing in as little as 3 hours. Compared to manual methods, the iBind western system requires less primary antibody, enables reproducible results, and is compatible with downstream detection protocols.

Each iBind PLUS Solution Kit contains sufficient reagents for 10 mini-size blot immunoprocessing runs.

For iBind Flex, use the iBind Flex PLUS Solution Kit. For fluorescent applications, use the iBind Fluorescent Detection (FD) Solution Kit (Cat. No. SLF1019) or iBind Flex Fluorescent Detection (FD) Solution Kit (Cat. No. SLF2019).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)iBind Western Device
Kit ContentsiBind PLUS Solution 250 mL, iBind PLUS Antibody Diluent, 45 mL
No. of Reactions10 Mini Blots
Quantity1 Kit
Detection MethodColorimetric, Chromogenic, Chemiluminescence
Gel CompatibilityMini Precast Protein Gels
Membrane CompatibilityNitrocellulose, PVDF
Product LineiBind
Product TypeWestern Blot Kit
Unit SizeEach
Contents & Storage
iBind PLUS Solution, iBind PLUS Antibody Diluent
Store at 2°C to 8°C