Lambda DNA/EcoRI plus HindIII Marker, ready-to-use
Thermo Scientific™
Lambda DNA/EcoRI plus HindIII Marker, ready-to-use
Thermo Scientific Lambda DNA/EcoRI+HindIII Marker, ready-to-use is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. LambdaRead more
Have Questions?
Catalog NumberQuantity
SM01935 x 50 μg
Catalog number SM0193
Price (MXN)
-
Quantity:
5 x 50 μg
Request bulk or custom format
Thermo Scientific Lambda DNA/EcoRI+HindIII Marker, ready-to-use is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
Lambda DNA/EcoRI+HindIII Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading (after heating) onto agarose gels.
Highlights
• Sizing and approximate quantification of large DNA fragments
• Sharp bands
• Premixed with 6X DNA Loading Dye for a direct loading (after heating) and for room temperature storage
• Supplied with loading dye for sample DNA
Note
The cohesive ends (the 12 nt cos site of lambda DNA) of fragments indicated may anneal and form additional bands. These fragments can be separated by heating at 65°C for 5 minutes and then cooling on ice for 3 minutes.
The shortest fragments (oblique) are not visible in standard electrophoresis. Fragment lengths are predicted by computer analysis of respective DNA sequences.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel CompatibilityAgarose Gels
Quantity5 x 50 μg
Ready to LoadYes
Gel TypeAgarose
Product TypeDNA/ECORI Plus Hind III Marker
Unit Size5 x 50 µg
Frequently asked questions (FAQs)
What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).