Ladders can now be stored at room temperature for up to 24 months! During transition period you may receive packing indicating 6-month RT stability.
O'RangeRuler DNA Ladders, ready-to-use
Thermo Scientific™

O'RangeRuler DNA Ladders, ready-to-use

Thermo Scientific O'RangeRuler DNA Ladders is a step ladder designed for precise sizing of DNA fragments in agarose or non-denaturing polyacrylamide gels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize Range
SM0653100 to 6000 bp
SM130310 to 100 bp
SM131310 to 150 bp
SM132320 to 300 bp
SM061350 to 1000 bp
SM0623100 to 500 bp
SM0633200 to 3000 bp
SM0643500 to 6000 bp
Catalog number SM0653
Price (USD)
132.00
25 µg
Size Range:
100 to 6000 bp
Request bulk or custom format
Price (USD)
132.00
25 µg

Thermo Scientific O'RangeRuler DNA Ladders are designed for precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels.

O'RangeRuler series is characterized by its 'step ladder' design. The chromatography-purified DNA fragments are precisely separated by a consistent increment (e.g., 5 bp, 10 bp, 20 bp, 50 bp, 100 bp, 200 bp, or 500 bp). The design enables accurate and precise sizing of DNA fragments. Not recommended for DNA quantification.

All O'RangeRuler DNA Ladders are supplied with 6X Orange DNA Loading Dye for sample DNA. Orange G dye can be used to monitor DNA migration in agarose or polyacrylamide gels and migrates at 50 bp in a 1% agarose gel.

Highlights

  • Step ladders available in a wide range of increments from 5-to-500 bp
  • Evenly spaced reference bands
  • Sharp bands, easy-to-remember fragment sizes
  • Ready-to-use—premixed with 6X Orange Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use O'RangeRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel CompatibilityAgarose Gels
Quantity25 μg
Ready to LoadYes
Shelf Life24 months when stored at room temperature.
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Orange G
Product LineO'GeneRuler, O’RangeRuler
Product TypeDNA Ladder
Size Range100 to 6000 bp
Unit Size25 µg

Frequently asked questions (FAQs)

Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.