Standard Sample Holder XL
Thermo Scientific™
Standard Sample Holder XL
The Standard Sample Holder for the Phenom XL Desktop SEM is the compact stage allowing analysis of samples of up to 100 mm x 100 mm. In spite of this much larger sample size, a proprietary loading shuttle keeps the vent/load cycle to a minimum, which in practice enables a throughput that is a few factors higher than any comparable SEM system.
Have Questions?
Catalog number STANDSAMPLEHOLD-XL
Price (USD)
-
The Thermo Scientific™ Standard Sample Holder for the Thermo Scientific™ Phenom™ XL Desktop SEM is the compact stage allowing analysis of samples of up to 100 mm x 100 mm. In spite of this much larger sample size, a proprietary loading shuttle keeps the vent/load cycle to a minimum, which in practice enables a throughput that is a few factors higher than any comparable SEM system.
Embedding, cutting and polishing are common techniques used to create flat samples for microscopic investigation. Typically these samples are embedded in a resin and can have a few standard size diameters.
The Standard Sample Holder for the Phenom XL can be extended with 3 types of resin or metallurgical mount inserts. This means that the sample holder can accommodate the most common sized resin mount samples.
Sample size
  • 100 mm x 100 mm (up to 36 x 12 mm pin stubs)
  • Max. 65 mm (h)