Orion Star™ A122 Conductivity Portable Meter
Thermo Scientific™

Orion Star™ A122 Conductivity Portable Meter

Take simple conductivity, TDS and temperature measurements with this conductivity portable meter with easy operation for basic field testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProbe Type
STARA12252-cell Conductivity Sensor
STARA1220Not included
Catalog number STARA1225
Price (USD)
1,377.00
Online exclusive
1,530.00
Save 153.00
Each
Probe Type:
2-cell Conductivity Sensor
Price (USD)
1,377.00
Online exclusive
1,530.00
Save 153.00
Each

Take simple, routine conductivity, total dissolved solids (TDS) and temperature measurements with the Thermo Scientific™ Orion Star™ A122 Conductivity Portable Meter. This instrument, intended for basic conductivity analysis in the field, is easy-to-use and budget-friendly. Obtain temperature compensated readings using linear option with 20°C or 25°C reference temperatures. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. This waterproof meter has an IP67-rated housing for rugged protection in outdoor environments.

Thermo Scientific Orion Star A122 conductivity portable meters are a simple, economical solution for your routine outdoor sample analysis. Featuring an emphasized measure key, selectable read modes and measurement stability indicator, our basic Orion Star A122 conductivity meters are designed to deliver measurements you can trust at an affordable price.

  • Measure conductivity or TDS with temperature
  • Perform a one point automatic or manual calibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range
  • Use linear temperature compensation with 20°C or 25°C reference temperatures
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
  • User-friendly operation with basic calibration text prompts
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications and 3-year replacement warranty

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Accuracy (Conductivity)0.5% of reading ±1 digit
Accuracy (TDS)0.5% of reading ±1 digit
Accuracy (Temperature)±0.1°C
Battery Life2000 hr.
Battery Type4 x AA (included)
CalibrationConductivity and temperature
Calibration Points (Conductivity)1
Cell Constant0.001 to 10cm-1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels1
Compatible Probes2-cell or 4-cell conductivity sensors with built-in temperature
Data Management DetailsInternal data point memory
DescriptionOrion Star A122 conductivity/TDS portable meter kit with 2-cell conductivity sensor, solutions, meter armor and field case
Dimensions (L x W x H)24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
IncludesStar A122 conductivity meter, 011050MD Orion 2-cell conductivity sensor, 011007 Orion 1413 µS conductivity standard, protective meter armor, hard-sided field case, 4 AA batteries, literature CD, printed quick start guide and meter test certificate
Inputs8 pin MiniDIN (conductivity sensor with built-in temperature)
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
Log Function TypeAutomatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A122 Conductivity/TDS/Temperature Portable Meter
OutputsNone
Power Supply4 AA batteries (included) or universal AC adapter (sold separately)
Probe Type2-cell Conductivity Sensor
Relative Humidity Range5 to 85%, non-condensing
Resolution (Conductivity)0.01 μS minimum, auto ranging up to 3 significant digits
Resolution (TDS)1 mg/L
Resolution (Temperature)0.1°C (0.1°F)
StabilitySmart Stability and Smart Averaging
TDS Factor RangeLinear (0.02 to 9.99)
Temperature CompensationLinear
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature (Metric) Reference20, 25°C
Temperature Probe Calibration1 point temperature offset calibration
Temperature SelectionManual or automatic with ATC temperature probe
TypePortable Conductivity Meter
Warranty3 Year Meter
Weight (English)2 lb.
Weight (Metric)0.5 kg
Data Points50
Display TypeLCD
IP RatingIP67
Measurement ModesConductivity, TDS with Temperature
Range (Conductivity)0.01 μS/cm to 200.0 mS/cm
Range (TDS)1 to 19,999 mg/L
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach

Frequently asked questions (FAQs)

How long is the warranty period for Orion conductivity meters?

At Thermo Fisher Scientific, we stand behind the quality and reliability of our instruments. That's why we offer a minimum two-year (2) warranty on all Orion benchtop and portable conductivity meters. Obtain peace of mind knowing that your investment is protected. Should any issues arise within the warranty period, our team of experts is committed to providing you with prompt and efficient service. Be sure to check instrument specifications to confirm the warranty coverage period.

How do I perform a calibration using Orion conductivity meters?

Calibration is an important step to obtaining accurate results on your Orion benchtop or portable conductivity meter. Before beginning the calibration process, ensure that you are utilizing freshly poured, non-expired calibration standards to ensure accurate results. Follow the steps below to calibrate your conductivity meter.

• Prepare the calibration standards: Obtain at least one conductivity reference standard with a known conductivity value near the expected sample conductivity range. Ideally, use one standard close to the expected conductivity of your sample, and additional standards for different conductivity ranges. Ensure that the standards are fresh and have not expired.
• Clean the probe: Rinse the probe with distilled or deionized water to remove any debris or contaminants. If necessary, gently wipe the probe with a soft cloth or tissue.
• Turn on the conductivity meter: Power on the conductivity meter and allow it to warm up, if needed. • Immerse the probe: Immerse the probe into a beaker containing the first calibration standard. Ensure that the probe is fully submerged and that it is not touching the sides or bottom of the beaker.
• Allow stabilization: Allow the conductivity reading to stabilize. This may take a few moments, depending on the specific meter and standard being used. The reading should become steady and remain constant.
• Adjust the conductivity value: Using the calibration controls on the conductivity meter, adjust the reading to match the known conductivity value of the calibration standard. Reference the user manual for specific guidance on adjusting the conductivity.
• Rinse the probe: After calibration, rinse the probe with distilled or deionized water to remove any residual calibration solution.
• Repeat calibration: Repeat the calibration steps with the second calibration standard, following the same process as before.
• Verify accuracy: After calibration, it is recommended to check the conductivity meter's accuracy by measuring a conductivity solution of known value. This can help ensure the reliability of your measurements.

To learn more about conductivity calibration, maintenance and beyond, download our user guide: Orion Conductivity Probe User Guide

How do I perform an analysis using Orion conductivity meters?

Before beginning an analysis on your Orion benchtop or portable conductivity meter, assemble your materials. You will need a conductivity probe suitable for your application, calibration standards, and a test sample. Follow the steps below to obtain your conductivity analysis.

• Calibrate the conductivity meter: Start by calibrating your conductivity meter using at least one calibration standard. Typically, you will need at least one standard with a known conductivity value near the expected sample conductivity range. Be sure to reference the user manual for instructions on how to properly calibrate your conductivity meter.
• Prepare your sample: Ensure that your sample is at the desired temperature (most frequently room temperature).
• Immerse the electrode: Dip the conductivity probe into the sample. Avoid touching the sides or bottom of the container to prevent inaccurate readings.
• Allow for stabilization: Give the conductivity meter some time to stabilize and reach a steady reading. This may take a few seconds to a minute depending on your meter and the characteristics of the sample.
• Record the conductivity value: Once the conductivity meter stabilizes, record the conductivity value and temperature value displayed on the meter. Take note of any temperature compensation settings if applicable. • Rinse the probe: After each measurement, rinse the probe with distilled or deionized water to remove any residue or contaminants. This ensures accurate subsequent readings.
• Repeat (if necessary): If you need to analyze multiple samples, repeat the process with each sample, recalibrating the conductivity meter if required.

To learn more about how to get accurate conductivity measurements, download the smart note: Top 10 Mistakes to Avoid When Measuring Conductivity in Liquids.

How do I choose an Orion conductivity probe for my application?

When choosing an Orion conductivity probe for your application there are many factors to consider. Measurement range, sample type, container dimensions, accuracy/ precision requirements, temperature compensation needs, cell constant, durability, maintenance, and meter compatibility are all key features to evaluate.

To learn more about Orion conductivity probes, download the brochure: Orion Conductivity Probes

What supplies do I need to perform an analysis with Orion conductivity meters?

To perform an analysis using Orion benchtop and portable conductivity meters, you will need the following supplies: conductivity probe, conductivity standards, distilled or deionized water, sample containers, and a stirrer probe (optional).