Orion Star™ A221 Portable pH Meter
Thermo Scientific™

Orion Star™ A221 Portable pH Meter

Reliably measure pH, mV, ORP and temperature with our portable pH meter for advanced field analysis and printer or computer interfacing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
STARA2216pH Meter Standard Kit
STARA2210
STARA2217pH Meter ORP Kit
STARA2218pH Meter Food/Beverage Kit
STARA2215
Catalog number STARA2216
Price (USD)
1,197.00
Online exclusive
1,330.00
Save 133.00
Each
Type:
pH Meter Standard Kit
Price (USD)
1,197.00
Online exclusive
1,330.00
Save 133.00
Each

Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A221 Portable pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the field. Perform up to a 5 point pH calibration, log up to 1000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Waterproof meter ensures durable, rugged performance in outdoor environments with an IP67-rated housing.

Don't leave your measurements to chance when analyzing samples outside the lab. Orion Star A221 Portable pH Meters are designed to offer the reliability you need in a user-friendly package for your dedicated pH testing needs. This waterproof, portable meter displays critical data with your measurement including temperature, date and time, read type, measurement stability, active calibration info and power source status.

  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 or ±0.05%, whichever is greater
Accuracy (pH)±0.002
Alarm OutputCalibration due alarm (fixed 12 hour)
Backlight OptionYes (Selectable)
Battery Life800 hr.
Battery Type4 x AA
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels1
Data Management DetailsTransfer single, range or all data points to printer or computer
DescriptionOrion Star A221 pH portable meter standard kit with general purpose low-maintenance pH/ATC electrode
Dimensions (L x W x H)24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
IP RatingIP67
IncludesStar A221 pH meter, 9107BNMD gel-filled epoxy-body pH/ATC electrode, pH 4/7/10 buffer and rinse pouches, storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable
InputsBNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Isopotential Point7.000pH
KeypadComprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
LanguagesEnglish, Spanish, French, Italian, German and Chinese; Portuguese and Korean Available with Software Update
MemoryNon-volatile memory preserves data log, calibration log and meter settings
ModelOrion Star A221 pH/mV/RmV/ORP/Temperature Portable Meter
OutputsUSB, RS-232
Power Supply4 AA batteries (included) or universal AC adapter (sold separately)
Probe TypepH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (includes 9107BNMD silver chloride pH/ATC electrode)
Range (ORP)±2000.0mV
Range (Relative mV)±2000.0mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (Temperature)0.1°C, 0.1°F
Resolution (mV)0.1mV
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
TypepH Meter Standard Kit
Weight (English)1.2 lb.
Weight (Metric)0.5 kg
CalibrationpH, relative mV (RmV), ORP and temperature
Calibration Points1 to 5
Data Points1000 with Date and Time Stamp
Display TypeLCD, Graphic with Backlight
Log Function TypeAutomatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
Measurement ModespH, mV, Relative mV, ORP with Temperature
Range (mV)±2000
Range (pH)-2 to 20
Resolution (pH)0.001, 0.01, 0.1
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach

Frequently asked questions (FAQs)

How long is the warranty period for Orion pH meters?

At Thermo Fisher Scientific, we stand behind the quality and reliability of our instruments. That's why we offer a minimum two-year (2) warranty on all Orion benchtop and portable pH meters. Obtain peace of mind knowing that your investment is protected. Should any issues arise within the warranty period, our team of experts is committed to providing you with prompt and efficient service. Be sure to check instrument specifications to confirm the warranty coverage period.

How do I perform a calibration using Orion pH meters?

Calibration is an important step to obtaining accurate results on your Orion benchtop or portable pH meter. Before beginning the calibration process, ensure that you are utilizing freshly poured, non-expired calibration buffers to help ensure accurate results. Follow the steps below to calibrate your pH meter.

• Prepare the calibration solutions: Obtain at least two pH reference solutions with known pH values. Ideally, use one solution close to the expected pH of your sample, and another solution at a different pH value. Ensure that the solutions are fresh and have not expired.
• Clean the electrode: Rinse the electrode with distilled or deionized water to remove any debris or contaminants. If necessary, gently wipe the electrode with a soft cloth or tissue.
• Turn on the pH meter: Power on the pH meter and allow it to warm up, if needed.
• Immerse the electrode: Immerse the electrode into a beaker containing the first calibration solution. Ensure that the electrode is fully submerged and that it is not touching the sides or bottom of the beaker.
• Allow stabilization: Allow the pH reading to stabilize. This may take a few moments, depending on the specific meter and solution being used. The reading should become steady and remain constant.
• Adjust the pH value: Using the calibration controls on the pH meter, adjust the reading to match the known pH value of the calibration solution. Reference the user manual for specific guidance on adjusting the pH.
• Rinse the electrode: After calibration, rinse the electrode with distilled or deionized water to remove any residual calibration solution.
• Repeat calibration: Repeat the calibration steps with the second calibration solution, following the same process as before.
• Verify accuracy: After calibration, it is recommended to check the pH meter's accuracy by measuring a pH solution of known value. This can help ensure the reliability of your measurements.

To learn more about pH meter calibration, download our technical note: Understanding Calibration of Laboratory pH Systems. See a live calibration by watching our video: Understanding Calibration of Laboratory pH Systems.

How do I perform an analysis using Orion pH meters?

Before beginning an analysis on your Orion benchtop or portable pH meter, assemble your materials. You will need a pH electrode suitable for your application, calibration buffer solutions, and a test sample. Follow the steps below to obtain your pH analysis.

• Calibrate the pH meter: Start by calibrating your pH meter using the calibration buffer solutions. Typically, you will need pH 4, pH 7, and pH 10. Be sure to reference the user manual for instructions on how to properly calibrate your pH meter.
• Prepare your sample: Ensure that your sample is at the desired temperature (most frequently room temperature). If necessary, filter the sample to remove any particles or impurities that could impact the pH reading.
• Immerse the electrode: Dip the pH electrode into the sample. Avoid touching the sides or bottom of the container to prevent inaccurate readings.
• Allow for stabilization: Give the pH meter some time to stabilize and reach a steady reading. This may take a few seconds to a minute depending on your meter and the characteristics of the sample.
• Record the pH value: Once the pH meter stabilizes, record the pH value displayed on the meter. If applicable, also record the temperature value.
• Rinse the electrode: After each measurement, rinse the electrode with distilled or deionized water to remove any residue or contaminants. This helps ensure accurate subsequent readings.
• Repeat (if necessary): If you need to analyze multiple samples, repeat the process with each sample, recalibrating the pH meter if required.

To learn more about how to perform a pH analysis, watch our video:Measuring Drinking Water Using the Orion Versa Star Pro Bench Meter

How do I choose an Orion pH electrode for my application?

When choosing an Orion pH electrode for your application there are many factors to consider. Sample type, container dimensions, accuracy/ precision requirements, junction design, durability, maintenance, and meter compatibility are all key features to evaluate.
To learn more about how to choose the right electrode for your application, watch our videos: How to Choose the Right pH Electrode and Understanding Your Sample to Select the Right pH Electrode

What supplies do I need to perform an analysis with Orion pH meters?

To perform an analysis using Orion benchtop and portable pH meters, you will need the following supplies: pH electrode (or micro electrode for small sample volumes/ containers), calibration buffer solutions, distilled or deionized water, sample containers, stirrer probe (optional), and electrode cleaning solution(s) (s). For your convenience, our all-in-one pH buffer kits can be purchased, which include pH buffers (4.01 pH, 7.00 pH, and 10.01 pH), electrode cleaning solution, and electrode and storage solution.