SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, nitrocellulose - FAQs
SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, nitrocellulose - FAQs
View additional product information for SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, nitrocellulose - FAQs (STM2007)
1 product FAQs found
Do you offer pre-cut membranes and filters for the SureLock Tandem Midi Blot Module?
We offer the following pre-cut membranes and filters for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, nitrocellulose, Cat. No. STM2008
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 µm, nitrocellulose, Cat. No. STM2007
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, PVDF, Cat. No. STM2006
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.