Galacto-Light Plus™ β-Galactosidase Reporter Gene Assay System
Invitrogen™

Galacto-Light Plus™ β-Galactosidase Reporter Gene Assay System

Rapid, ultrasensitive detection of β-galactosidase in reporter gene assays. Thermo Scientific Galacto-Light Plus β-Galactosidase Reporter Gene Assay System is designed for luminometers without injectors.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
T1007200 Assays
T100915,000 Assays
T1011600 Assays
Catalog number T1007
Price (USD)
447.00
200 assays
Quantity:
200 Assays
Request bulk or custom format
Price (USD)
447.00
200 assays

The Galacto-Light Plus™ Beta-Galactosidase Reporter Gene Assay System is designed for the rapid, ultrasensitive detection of β-galactosidase in reporter gene assays.

  • The Galacto-Light Plus™ Beta-Galactosidase Reporter Gene Assay System is designed for luminometers without injectors.
  • Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range.
  • Assay sensitivity is 100- to 1,000-fold better than with either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays.
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions.
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes.
  • Assay can be completed in about one hour, providing fast assay turnaround.

Sensitive

This chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.

Ready-to-Use

The Galacto-Light™ Plus™ system incorporates chemiluminescent substrates, reaction buffer, and light-emission accelerator containing luminescence enhancers, in a ready-to-use format. The reaction buffer is supplied at a pH that aids in discrimination of endogenous β-galactosidase, while having little effect on the bacterial form of the enzyme. This enables sensitive detection even in cell lines with relatively high levels of endogenous β-galactosidase activity.

Products described herein are guaranteed for 12 months from the date of purchase.

For Research Use Only. Not for use in diagnostics procedures.

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assayβ-Gal (lacZ) Assay
Compatible CellsMammalian Cells
Detection MethodChemiluminescence
For Use With (Application)Enzyme Assay
For Use With (Equipment)Luminometer
FormLiquid
High-throughput CompatibilityHigh-throughput Compatible
Label or DyeUnconjugated
Product LineGalacto-Light Plus, NovaBright
Product Typeβ-Galactosidase Reporter Gene Assay System
Quantity200 Assays
ReadoutEnd Point
Sample TypeCell Cultures
SubstrateGalacton-Plus™
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Gal Substrate
Target EnzymeBeta-Galactosidase
Unit Size200 assays
Contents & Storage
This kit contains the lysis solution, substrate, reaction buffer, and accelerator.
Store at 4°C.