ThiolTracker™ Violet (glutathione detection reagent) - FAQs
ThiolTracker™ Violet (glutathione detection reagent) - FAQs
View additional product information for ThiolTracker™ Violet (glutathione detection reagent) - FAQs (T10095)
2 product FAQs found
Is ThiolTracker Violet a fluorogenic reagent?
No. ThiolTracker Violet is fluorescent whether bound or unbound to a sulfhydryl group.
Will ThiolTracker Violet bind to proteins as well as glutathione?
Yes, ThiolTracker Violet will bind to any accessible sulfhydryl group (-SH; cysteine) whether it is on reduced glutathione or on the surface of a protein. It will not bind to sulfur bound via a disulfide bond (cystine) or ‘buried’ cysteines.