Galacto-<i>Star</i>&trade; &beta;-Galactosidase Reporter Gene Assay System for Mammalian Cells
Invitrogen™

Galacto-Star™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells

The Galacto-Star™ One-Step β-Galactosidase Reporter Gene Assay System chemiluminescent reporter gene assay system enables rapid, sensitive detection of β-galactosidase in mammalian cell lysates. SensitiveThe chemiluminescent assay exhibits over three o

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
T10141800 Assays
T1012600 Assays
Catalog number T1014
Price (USD)
1,030.00
1,800 assays
Quantity:
1800 Assays
Request bulk or custom format
Price (USD)
1,030.00
1,800 assays

The Galacto-Star™ One-Step β-galactosidase Reporter Gene Assay System chemiluminescent reporter gene assay system enables rapid, sensitive detection of β-galactosidase in mammalian cell lysates.

  • Glow assay kinetics provides a 'window' during which measurements may be performed, facilitating HTS applications where assay automation is used.
  • Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems.
  • Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range.
  • Assay sensitivity is 100- to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays.
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions.
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes.
  • Assay can be completed in about one hour, providing fast assay turnaround.

Sensitive

The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity than colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.

Versatile

The Galacto-Star™ system includes Galacton-Star™ chemiluminescent substrate for β-galactosidase and is suited for measurement in both luminometers and scintillation counters. Automatic injection is not required.

Long-Lasting Signal

Light emission from the Galacto-Star™ assay reaches maximum in 60-90 minutes and remains constant for nearly one hour. Cell lysate is mixed with reaction buffer containing Galacton-Star™ substrate and Sapphire-II™ luminescence enhancer. After a 60-90 minute incubation at room temperature, the signal is measured in a luminometer or scintillation counter.

For Research Use Only. Not for use in diagnostics procedures.

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assayβ-Gal (lacZ) Assay
Compatible CellsMammalian Cells
Detection MethodChemiluminescence
For Use With (Application)Enzyme Assay
For Use With (Equipment)Luminometer
FormLiquid
High-throughput CompatibilityHigh-throughput Compatible
Label or DyeUnconjugated
Product LineGalacto-Star, NovaBright
Product Typeβ-Galactosidase Reporter Gene Assay System
Quantity1800 Assays
ReadoutEnd Point
Sample TypeCell Cultures
SubstrateGalacton-Star™
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Gal Substrate
Target EnzymeBeta-Galactosidase
Unit Size1,800 assays
Contents & Storage
This kit contains the lysis solution, substrate, enhancer, and required buffer. Store the product at 4°C.