Phospha-Light™ SEAP Reporter Gene Assay System, 1200 Assays - FAQs
Phospha-Light™ SEAP Reporter Gene Assay System, 1200 Assays - FAQs
View additional product information for Phospha-Light™ SEAP Reporter Gene Assay System - FAQs (T1015, T1016, T1017)
1 product FAQs found
Do you offer an alternative for SEAP Substrate Luminescence (Cat. No. A63880)?
We suggest the following products as an alternative to for SEAP Substrate Luminescence (Cat. No. A63880):
Phospha-Light SEAP Reporter Gene Assay System (Cat. Nos. T1015, T1016, T1017)
NovaBright Phospha-Light EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection (Cat. Nos. N10577, N10578)
Both kits utilize the high-performance alkaline phosphatase substrate, CSPD, and an Emerald Enhancer.