Galacto-Light™ Reaction Buffer Diluent with Galacton-Plus™ Substrate
Invitrogen™

Galacto-Light™ Reaction Buffer Diluent with Galacton-Plus™ Substrate

Established system with extended light emission and high sensitivity for reporter gene assays. Thermo Scientific Galacto-Light Reaction Buffer Diluent and the Galacton-Plus chemiluminescent substrate solution are used with the Galacto-Light Plus Reporter

Have Questions?
Catalog NumberQuantity
T10551800 assays
Catalog number T1055
Price (USD)
381.00
1800 assays
Quantity:
1800 assays
Request bulk or custom format
Price (USD)
381.00
1800 assays

The Galacto-Light™ Reaction Buffer Diluent and the Galacton-Plus™ chemiluminescent substrate solution are used with the Galacto-Light™ Plus Reporter Gene Assay System.

Use With

Light Emission Accelerator-II

For Research Use Only. Not for use in diagnostics procedures.

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateUnconjugated
Detection MethodChemiluminescence
For Use With (Application)Enzyme Assay
For Use With (Equipment)Luminometer (Microplate)
FormLiquid
Format96-well Plates
High-throughput CompatibilityHigh-throughput Compatible
Product LineGalacto-Light, Galacton-Plus
Sample TypeCell Cultures
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Gal
Product TypeELISA Substrate and Diluent
Quantity1800 assays
Unit Size1800 assays
Contents & Storage
This kit contains the concentrated substrate and the dilution buffer.
Store the product at 4°C.