CSPD™ Substrate (0.4 mM Ready-To-Use) with Sapphire-II™ Enhancer, Size B
Invitrogen™

CSPD™ Substrate (0.4 mM Ready-To-Use) with Sapphire-II™ Enhancer, Size B

This CSPD™ substrate with Sapphire-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting onRead more
Have Questions?
Catalog NumberQuantity
T2210100 ml
Catalog number T2210
Price (USD)
866.40
Special offer
Online exclusive
Ends: 27-Jun-2026
912.00
Save 45.60 (5%)
Each
Quantity:
100 ml
Request bulk or custom format
Price (USD)
866.40
Special offer
Online exclusive
Ends: 27-Jun-2026
912.00
Save 45.60 (5%)
Each
This CSPD™ substrate with Sapphire-II™ Enhancer is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

• Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
• Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours. Solution assays require chemiluminescent enhancers. Membrane-based assays may also require them for increased light output and sensitivity.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodChemiluminescence
FormLiquid
FormatBottle
Product LineCSPD, NovaBright
Shipping ConditionRoom Temperature
Substrate TypeAlkaline Phosphatase
Product TypeELISA Substrate
Quantity100 ml
Unit SizeEach
Contents & Storage
Store the product at 4°C.