With Tubulin Tracker Deep Red and Tubulin Tracker Green, can I fix samples after labeling?
No. The labeling is not retained upon fixation. Imaging of the samples should be done only on live cells. To limit photobleaching while viewing live cells, use ProLong Live Antifade Reagent, for live cell imaging (Cat No. P36974 and P36975).