The isomeric mixture of amine-reactive Texas Red-X, succinimidyl ester and its conjugates emit at a longer wavelength than do eitherRead more
Have Questions?
Catalog Number
Quantity
T6134
5 mg
Catalog number T6134
Price (USD)
452.00
5 mg
Quantity:
5 mg
Price (USD)
452.00
5 mg
The isomeric mixture of amine-reactive Texas Red-X, succinimidyl ester and its conjugates emit at a longer wavelength than do either tetramethylrhodamine or Lissamine rhodamine B, and are among the most commonly used long-wavelength “third labels” in fluorescence microscopy and flow cytometry. Protein conjugates prepared with Texas Red-X succinimidyl ester also have a higher fluorescence yield than those with the same labeling ratio prepared with Texas Red sulfonyl chloride.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Label or DyeTexas Red™
Product TypeSuccinimidyl Ester
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Product LineTexas Red
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (39)
Citations & References
Abstract
A lipid receptor sorts polyomavirus from the endolysosome to the endoplasmic reticulum to cause infection.
Authors:Qian M, Cai D, Verhey KJ, Tsai B,
Journal:PLoS Pathog
PubMed ID:19503604
'The mechanisms by which receptors guide intracellular virus transport are poorly characterized. The murine polyomavirus (Py) binds to the lipid receptor ganglioside GD1a and traffics to the endoplasmic reticulum (ER) where it enters the cytosol and then the nucleus to initiate infection. How Py reaches the ER is unclear. We ... More
TRPV1 regulators mediate gentamicin penetration of cultured kidney cells.
Authors:Myrdal SE, Steyger PS
Journal:Hear Res
PubMed ID:15925202
'Transient receptor potential (TRP) receptors are, typically, calcium-permeant cation channels that transduce environmental stimuli. Both kidney epithelial and inner ear sensory cells express TRPV1, are mechanosensors and accumulate the aminoglycoside antibiotic gentamicin. Recently, we showed that Texas Red-conjugated gentamicin (GTTR) enters kidney cells via an endosome-independent pathway. Here, we used ... More
Cytoplasmic and intra-nuclear binding of gentamicin does not require endocytosis.
Authors:Myrdal SE, Johnson KC, Steyger PS
Journal:Hear Res
PubMed ID:15925201
'Understanding the cellular mechanism(s) by which the oto- and nephrotoxic aminoglycoside antibiotics penetrate cells, and the precise intracellular distribution of these molecules, will enable identification of aminoglycoside-sensitive targets, and potential uptake blockers. Clones of two kidney cell lines, OK and MDCK, were treated with the aminoglycoside gentamicin linked to the ... More
Interaction of Mycobacterium avium-containing phagosomes with the antigen presentation pathway.
Authors:Ullrich HJ, Beatty WL, Russell DG
Journal:J Immunol
PubMed ID:11086039
'Pathogenic mycobacteria infect macrophages where they replicate in phagosomes that minimize contact with late endosomal/lysosomal compartments. Loading of Ags to MHC class II molecules occurs in specialized compartments with late endosomal characteristics. This points to a sequestration of mycobacteria-containing phagosomes from the sites where Ags meet MHC class II molecules. ... More
Evolution of peptides that modulate the spectral qualities of bound, small-molecule fluorophores.
Authors:Rozinov MN, Nolan GP
Journal:Chem Biol
PubMed ID:9862799
'BACKGROUND: Fluorophore dyes are used extensively in biomedical research to sensitively assay cellular constituents and physiology. We have created, as proof of principle, fluorophore dye binding peptides that could have applications in fluorescent dye-based approaches in vitro and in vivo. RESULTS: A panel of Texas red, Rhodamine red, Oregon green ... More