TDE Series -86°C Ultra-Low Temperature Chest Freezers - FAQs
TDE Series -86°C Ultra-Low Temperature Chest Freezers - FAQs
View additional product information for TDE Series -86°C Ultra-Low Temperature Chest Freezers, Medical Device - FAQs (TDEC06386LV, TDEC25286LA, TDEC06386LA, TDEC25286LD, TDEC39686LA, TDEC39686LV, TDEC25286LV, TDEC39686LD)