TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometers
TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometers
TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometers
TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometers
TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometers
Thermo Scientific™
TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometers
Achieve next-level productivity, versatility, performance, and simplicity for high-confidence, high-throughput, targeted screening, and quantitation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeACTLabel
TSQ03-10001TSQ LC-MS with Oil Pump
TSQ03-11001TSQ LC-MS with Dry Pump
Catalog number TSQ03-10001
Price (USD)
-
Type:
TSQ LC-MS with Oil Pump
Achieve next-level productivity, versatility, and performance for method development and routine sample analysis. The TSQ Quantis Plus triple quadrupole mass spectrometer combines enhanced acquisition speed and sensitivity with proven robustness and reliability to deliver confident targeted screening and precise quantitation for high-throughput applications. Advanced instrument control software, compound database integration, and predefined method templates simplify operation, enabling LC-MS/MS users of all skill levels to obtain high-confidence results.
The TSQ Quantis Plus mass spectrometer seamlessly integrates with industry-leading quantitation software, (U)HPLC and HPIC systems, and sample preparation solutions; it also offers enhanced efficiency for targeted screening and quantitation workflows for a wide array of molecular compounds. Other benefits include:
  • Superior SRM acquisition speed to manage large-scale experimental studies with high-throughput requirements.
  • Outstanding robustness, enabling increased confidence in data acquisition with prolonged instrument uptime
  • Confident quantitation covering six orders of dynamic range
  • Simplicity and ease-of-use, allowing operators of all expertise levels to acquire high-quality data with improved confidence in results
  • Providing direct method transfer from the current TSQ Quantis mass spectrometer to support scalability
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only. Not For Diagnostic Procedures.
Specifications
Acquisition Speed800 (SRMs/sec)
Dimensions (D x W x H)33 x 30 x 27 in. (840 x 760 x 680 mm)
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, Forensic Toxicology
For Use With (Equipment)OptaMax NG ion source, FAIMS Pro Duo interface, VeriSpray PaperSpray ion source, EASY-Spray Series ion source, Nanospray Flex Series ion source
Polarity Switching5 (msec)
Software CompatibilityChromeleon CDS, TraceFinder software, Xcalibur software, FreeStyle software and mzCloud database
Mass Range2 to 3000 m/z
Resolution0.4 FWHM
TypeTSQ LC-MS with Oil Pump
Unit SizeEach