Vybrant™ Cell Adhesion Assay Kit - FAQs
Vybrant™ Cell Adhesion Assay Kit - FAQs
View additional product information for Vybrant™ Cell Adhesion Assay Kit - FAQs (V13181)
1 product FAQs found
What are the fluorescence excitation/emission maxima for the Calcein AM and SYTOX Green nucleic acid stain provided in the Vybrant Cell Adhesion Assay Kit?
Calcein AM: Ex/Em = 494 nm/517 nm
SYTOX Green nucleic acid stain: Ex/Em = 504 nm/523 nm