When using Dead Cell Apoptosis Kits with Annexin V for Flow Cytometry, what does a PI+ only population (hence Annexin V negative) correspond to?
These products are cells that died without undergoing apoptosis. A PI+ only population could be either a result of experimental condition or sample preparation.