CyQUANT™ Cytotoxicity Assay Kit (G6PD Release Assay) - FAQs
CyQUANT™ Cytotoxicity Assay Kit (G6PD Release Assay) - FAQs
View additional product information for CyQUANT™ Cytotoxicity Assay Kit (G6PD Release Assay) - FAQs (V23111)
3 product FAQs found
What are the fluorescence excitation/emission maxima for Resorufin that is generated in the Vybrant Cytotoxicity Assay Kit (G6PD Release Assay)?
The excitation/emission maxima for Resorufin are 563/587 nm.
How many assays can I perform using the CyQUANT Cytotoxicity Assay Kit (G6PD Release Assay) (Cat. No. V23111)?
The CyQUANT Cytotoxicity Assay Kit (G6PD Release Assay) (Cat. No. V23111) contains sufficient reagents to perform 1,000 assays using 100 µL reaction volumes.
Can I use flash-frozen supernatant from the cell culture for the CyQUANT Cytotoxicity Assay (Cat. No. V23111)?
Using flash‐frozen supernatant is not recommended for the CyQUANT Cytotoxicity Assay. It is better to take the reading straight after addition of the cytotoxic agent followed by an incubation and addition of the 2X Resazurin/Reaction Mixture.
Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center