What are the fluorescence excitation/emission maxima for the FAM-DEVD-FMK FLICA reagent, Hoechst 33342, and propidium iodide supplied in the Vybrant FAM Caspase-3 and -7 Assay Kit, for flow cytometry?
FAM-DEVD-FMK FLICA reagent: 488/530 nm
Hoechst 33342: 350/461 nm
Propidium iodide: 535/617 nm
Hoechst 33342: 350/461 nm
Propidium iodide: 535/617 nm