Product Image
Invitrogen™

Vivid Colors™ pcDNA™6.2/C-EmGFP-DEST Vector

The Vivid Colors™ Fluorescent Protein Gateway™ Destination Vectors allow you to quickly and easily fuse a protein of interest toRead more
Have Questions?
Catalog NumberQuantity
V35520
also known as V355-20
6 μg
Catalog number V35520
also known as V355-20
Price (USD)
2,800.00
6 µg
Quantity:
6 μg
Price (USD)
2,800.00
6 µg
The Vivid Colors™ Fluorescent Protein Gateway™ Destination Vectors allow you to quickly and easily fuse a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria (1,2) using the pcDNA™ 6.2 Gateway™ mammalian expression vector. These powerful Gateway™ Technology vectors contain the next-generation EGFP, Emerald Green Fluorescent Protein (EmGFP), or the popular Yellow Fluorescent Protein (YFP) for simple, non-invasive detection of recombinant protein. Both fluorescent proteins have been humanized for optimal mammalian expression (3). In addition, the Vivid Colors™ pcDNA™ 6.2 Fluorescent Protein Gateway™ Vectors offer:

• CMV promoter for high-level expression of the recombinant fluorescent fusion protein
• Options to fuse EmGFP or YFP to the N- and C-terminus of your protein
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
For Use With (Application)Reporter Assays, Subcellular Localization
Product TypeMammalian Expression Vector
Quantity6 μg
Reporter GeneGFP (EmGFP)
Selection Agent (Eukaryotic)Blasticidin
VectorpDEST, pcDNA
Cloning MethodGateway
Product LineGateway, Vivid Colors, pcDNA
PromoterCMV
Protein TagGFP (EmGFP)
Unit Size6 µg
Contents & Storage
Each vector is supplied as supercoiled and lyophilized plasmid DNA. Store at -20°C after resuspended. Guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

Can GFP fluorescence be detected in cells that have been stained for beta-galactosidase?

We recommend looking for GFP fluorescence before staining for beta-galactosidase. This is because the beta-galactosidase staining process produces a very high autofluorescence that will interfere with detection of GFP fluorescence.

What are the recommended filter sets for detection of EmGFP, YFP, CFP, and BFP by fluorescence microscopy?

EmGFP, YFP, CFP, and BFP can be detected using standard FITC filter sets and settings. However, for optimal detection of the fluorescence signal, filter sets optimized for detection within the excitation and emission ranges for each fluorescent protein are recommended. The recommended filter sets are as follows: EmGFP: Omega filter set XF100 YFP: Omega filter set XF1042 Chroma filter set 41028 CFP: Omega filter set XF114 Chroma filter set 31044 BFP: Omega filter set XF10 Chroma filter set 31021 For information on obtaining filter sets, please contact Omega Optical, Inc. (www.omegafilters.com) or Chroma Technology Corporation (www.chroma.com) directly.

What are the excitation and emission maxima for your fluorescent proteins (EmGFP, YFP, BFP, CFP, and Cycle 3 GFP)?

Excitation and emission maxima for our fluorescent proteins are as follows:
- EmGFP: Excitation: 487 nm; Emission: 509 nm
- YFP: Excitation: 514 nm; Emission: 527 nm
- BFP: Excitation: 308-383 nm; Emission: 440-447 nm
- CFP: Excitation: 452 nm; Emission: 505 nm
- Cycle 3 GFP: Primary excitation: 395 nm; Secondary Excitation: 478 nm; Emission: 507 nm

Are the fluorescent proteins you offer (EmGFP, YFP, CFP, BFP, and Cycle 3 GFP) humanized?

Yes, all of the fluorescent proteins offered by us (EmGFP, YFP, CFP, BFP, and Cycle 3 GFP) have been humanized for optimal mammalian expression.

How does Cycle 3 GFP compare with EmGFP and EGFP?

EmGFP is the next-generation variant of EGFP, and it has been further optimized for mammalian expression. Both EmGFP and EGFP can be visualized using the same filter sets (FITC) and settings. When used with the recommended filter sets and settings, Cycle 3 GFP is as bright as EGFP or EmGFP. However, when used with FITC filter sets and settings, Cycle 3 GFP is not as bright as EmGFP or EGFP.

- Excitation/emission maxima for EGFP: 488 nm/507-509 nm
- Excitation/emission maxima for EmGFP: 487 nm/509 nm
- Excitation/emission maxima for Cycle 3 GFP: 395 nm (primary) and 478 nm (secondary)/507 nm