BsdCassette™ Vector (pCMV/Bsd) - FAQs
BsdCassette™ Vector (pCMV/Bsd) - FAQs
View additional product information for BsdCassette™ Vector (pCMV/Bsd) - FAQs (V51020)
3 product FAQs found
I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?
Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.
Are your vectors routinely sequenced?
No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.
How was the reference sequence for your vectors created?
Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.