Versa Frame 44 Heavy Duty Checkweigher - FAQs
Versa Frame 44 Heavy Duty Checkweigher - FAQs
View additional product information for Versa Frame 44 Heavy Duty Checkweigher - FAQs (VERSAFRAME44)
7 product FAQs found
Can an existing Frame 44 checkweigher system be upgraded?
Yes. Facilities operating legacy Frame 44HB scales can extend the life of their equipment with a Versa Conversion Kit, which upgrades the controller to the latest Versa platform while retaining the existing weigh conveyor when appropriate.
How does the Versa Frame 44 checkweigher integrate with warehouse automation systems?
The Thermo Scientific Versa Frame 44 Heavy Duty Checkweigher system supports Ethernet and serial communications for weight output and includes Modbus connectivity for status and control. These capabilities allow seamless integration with warehouse management systems (WMS), shipping software, and automated material handling systems.
Is the Versa Frame 44 checkweigher suitable for harsh industrial environments?
Yes. The scale on the Thermo Scientific Versa Frame 44 Heavy Duty Checkweigher features a stainless-steel frame, heavy-duty load cells, and rugged construction designed for demanding warehouse and industrial applications. An optional IP65 washdown package is available for environments that require frequent cleaning or exposure to moisture.
Can the Versa Frame 44 checkweigher accurately weigh packages of different sizes?
Yes. Versa Warehouse software automatically adjusts to varying package dimensions and conveyor speeds. The Thermo Scientific Versa Frame 44 Heavy Duty Checkweigher can accurately weigh packages of different lengths without requiring product classification, reference weights, or a product database.
How fast can the Versa Frame 44 operate?
The Thermo Scientific Versa Frame 44 Heavy Duty Checkweigher is designed for high-throughput operations, supporting line rates up to 100 packages per minute. The Versa Frame 44XD model can operate at conveyor speeds up to 600 feet per minute, making it ideal for busy distribution centers and parcel handling facilities.
What is the maximum weighing capacity of the Versa Frame 44 checkweigher?
The Thermo Scientific Versa Frame 44HD Checkweigher supports package weights up to 100 lbs (45 kg), while the Versa Frame 44XD is available for applications requiring capacities up to 250 lbs (113 kg). This flexibility allows facilities to select the model that best fits their operational requirements.
What types of packages can the Versa Frame 44 checkweigher weigh?
The Thermo Scientific Versa Frame 44 Heavy Duty Checkweigher is designed for accurate in-line weighing of mailers, totes, cartons, cases, bags, and boxes in warehouse, distribution, and logistics environments. Its rugged construction makes it suitable for handling large and heavy packages moving through automated conveyor systems.