Integrated Gas Module for Varioskan™ LUX, Integrated gas module (factory installed at instrument purchase) - FAQs
Integrated Gas Module for Varioskan™ LUX, Integrated gas module (factory installed at instrument purchase) - FAQs
View additional product information for Integrated Gas Module for Varioskan™ LUX - FAQs (VLGAS001)
1 product FAQs found
Can I add gas control after purchase of the Varioskan LUX Multimode Microplate Reader?
No, the gas control module must be included with the instrument at the time of purchase. Please contact your Thermo Fisher Scientific sales representative for further information.