Multi-Attribute Method 2.0
Actual product may vary
Thermo Scientific™
Multi-Attribute Method 2.0
Use our Thermo Scientific™ Multi-Attribute Method (MAM) 2.0 workflows for an end-to-end, single-vendor solution that provides a holistic, compliance-ready platform for deployment from development to quality control.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLiquid Chromatography SystemMass Spectrometer Interface
WFLOW-64005Horizon Binary UHPLC SystemExploris MX Mass Detector
WFLOW-64002Flex Binary UHPLC SystemExploris 240 Mass Spectrometer
WFLOW-64003Horizon Binary UHPLC SystemExploris 240 Mass Spectrometer
WFLOW-64004Flex Binary UHPLC SystemExploris MX Mass Detector
Catalog number WFLOW-64005
Price (USD)
-
Liquid Chromatography System:
Horizon Binary UHPLC System
Mass Spectrometer Interface:
Exploris MX Mass Detector
Experience an end-to-end, single-vendor solution that provides a holistic, compliance-ready platform for deployment from development to quality control with our Thermo Scientific™ Multi-Attribute Method (MAM) 2.0 workflows. Developed through close partnership with the biopharmaceutical industry, MAM 2.0 enables:
  • Fast decision-making based on high confidence information of multiple product quality attributes (PQAs) from development through QC, ensured by industry-proven Thermo Scientific™ Orbitrap™ technology and the Chromeleon™ CDS as GMP supporting data acquisition and processing software.
  • Seamless knowledge sharing and method transfer across instruments, functions, departments, and sites throughout a whole organization to accelerate development, enabled by a compliance-ready enterprise data system.
  • Maximum productivity through a dedicated global support team of Thermo Fisher MAM experts, providing application specific service, support, and training.
Key Features of MAM 2.0
  • Enterprise connectivity for enhanced knowledge sharing
    Build your process knowledge within a compliance-ready, enterprise MAM data system that connects instruments, functions, departments, and sites throughout development and manufacturing, enabling seamless data and knowledge sharing.
  • Improve the efficiency of your biotherapeutic development and QC with industry proven Orbitrap technology, now purposely designed for MAM
    Simplify your routine operations without compromising data quality with the Orbitrap™ Exploris™ MX mass detector, built on the same platform as Orbitrap™ Exploris™ 240 mass spectrometer. Eliminate lengthy method development in QC with direct method transfer within compliance-ready Chromeleon™ CDS software.
  • Maximize productivity and confidence with our dedicated MAM Support Team
    A combination of on-site and remote trainings are conducted by MAM application experts, customizable to meet your needs. Installation, training, service, and support are managed by a dedicated global support team of Thermo Scientific MAM experts, providing fast responses to maximize system uptime and your productivity.
The Thermo Scientific Orbitrap Exploris and Excedion series mass spectrometers are Orbitrap instruments with an atmospheric pressure ionization (API) source for liquid chromatography (LC) mass spectrometry (MS) high-throughput applications. The instruments are designed to be placed on a bench in the laboratory and maintain mass accuracy across chromatographic peaks, supporting confident identification and quantitation using high-resolution accurate-mass detection.
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only. Not For Diagnostic Procedures.
Specifications
For Use With (Application)UHPLC, HRAM MS
IncludesUser guide, training, System Performance Evaluation Tests, service and support by a global, trained and dedicated team of Thermo Scientific MAM experts
Liquid Chromatography SystemHorizon Binary UHPLC System
Mass Spectrometer InterfaceExploris MX Mass Detector
SoftwareThermo Scientific™ Chromeleon™ CDS 7.3.1
TypeMulti-attribute Method
Unit SizeEach