Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Mouse) - FAQs
Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Mouse) - FAQs
View additional product information for Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Mouse) - FAQs (WP20006)
2 product FAQs found
Can I purchase the Secondary Antibody solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?
The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007), but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I purchase the Secondary Antibody solutions from the WesternBreeze Chromogenic Detection kits as standalone products?
The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007) but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.