New Zenon Alexa Fluor Plus Labeling Reagents are superior in three ways: improved bioconjugation for brighter signal, redesigned protein for robust screening regardless of IgG isotype, and new, more sensitive fluorophores.
Zenon&trade; Mouse IgG<sub>2a</sub> Labeling Kits
Invitrogen™

Zenon™ Mouse IgG2a Labeling Kits

Quickly generate antibody isotype conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ mouse IgG2a Labeling Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityExcitation/EmissionLabel or Dye
Z2515125 Reactions kit650/660 nmAPC (Allophycocyanin)
Z2510250 Reactions kit495/519 nmAlexa Fluor 488
Z2510750 Reactions kit590/617 nmAlexa Fluor 594
Z2510850 Reactions kit650/668 nmAlexa Fluor 647
Z2515525 Reactions kit496, 546, 565/578 nmR-PE (R-Phycoerythrin)
Catalog number Z25151
Price (USD)
409.65
Online exclusive
546.00
Save 136.35
1 kit
Quantity:
25 Reactions kit
Excitation/Emission:
650/660 nm
Label or Dye:
APC (Allophycocyanin)
Price (USD)
409.65
Online exclusive
546.00
Save 136.35
1 kit
Quickly generate antibody isotype conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ mouse IgG2a Labeling Kits. Simplify your laboratory applications and reduce antibody cross-reactivity while also achieving efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Achieve fast, versatile, and reliable fluorophore-, biotin-, or enzyme-labeled IgG2a primary antibodies with the Zenon™ mouse IgG2a labeling kits. These kits utilize Alexa Fluor fluorophores, biotin, Pacific Blue, or enzymes such as R-phycoerythrin and allophycocyanin, which are attached to monovalent, affinity purified Fab fragments. The Fab fragments, in turn, are directed against and bind with the Fc portion of IgG2a primary antibodies. Only a small amount of starting material is required, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Because the Zenon labeling method is based on immunoselectivity, it does not require the removal of exogenous proteins (such as serum) or amine-containing buffers from the target antibody, simplifying the process.

Zenon labeling technology greatly simplifies the use of multiple mouse-derived antibodies in the same staining protocol. Zenon tricolor labeling kits contain sufficient materials for 10 labeling reactions of each of three different fluorescent colors.

Important features of Zenon labeling technology:
• Labeled antibodies are typically ready to use in 10 minutes
• Requires only 1–20 μg primary antibody
• Simple—no purification required
• Flexible—choose from different fluorophores, biotin, HRP, alkaline phosphatase
• Multiplex with other mouse monoclonal antibodies simultaneously
• Can be used in a variety of applications including ICC, IHC, flow cytometry, and cell imaging.

Advantages of using Zenon antibody labeling kits:

Cost savings
Zenon antibody labeling kits offer a cost-conscious and reproducible method of tagging as little as 0.4 μg in 2 μL of primary antibody, with minimal waste of expensive or difficult-to-obtain antibodies, or excessive washing steps that pose the risk of product loss.

Sensitivity
Label your primary antibodies without compromising their antigen binding affinity: Zenon dye- and enzyme-labeled Fab fragments, which are targeted to the Fc tail, are affinity purified during their preparation to ensure high affinity and selectivity for the Fc portion of the corresponding primary antibody. The procedure for chemical labeling of the Zenon Fab fragments protects their Fc-binding site, resulting in more active labeling reagents.

Speed
No purification procedure is required prior to using Zenon Fab fragments in your laboratory applications. Formation of the Fab-antibody complex occurs in fewer than five minutes, followed by a five-minute blocking step. During this time, almost all the primary antibody in the mixture is labeled with the labeled Fab fragments.

Simplicity
The Fab-antibody complexes display fluorescence or enzymatic activity that is similar in intensity to that of directly labeled primary antibodies. Varying the extent of the antibody labeling is as simple as changing the amount of added Zenon labeling reagent during the reaction. Once the labeling complexes are formed, they can used immediately, without need for antibody purification.

Reliability
The Zenon Fab-antibody complex is stable and allows subsequent or simultaneous labeling of different target cells and tissues with different complexes. After staining, an aldehyde-based fixing step may be used to prevent the transfer of different labels between different primary antibodies, preserving the initial staining pattern.

Customization
We offer custom antibody conjugation services that are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
Excitation/Emission650/660 nm
Label TypePE & APC
Labeling Scale< 1–20 μg
Product LineZenon
Quantity25 Reactions kit
SpeciesMouse
Labeling TargetIgG2a
Label or DyeAPC (Allophycocyanin)
Unit Size1 kit
Contents & Storage
Contains 1 vial of Zenon APC mouse IgG2a labeling reagent (125 μL), and 1 vial of Zenon blocking reagent (mouse IgG, 125 μL).

Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

How should I store my allophycocyanin (APC)-conjugated antibody?

Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.