Zenon™ Allophycocyanin Mouse IgG2a Labeling Kit, 25 Reactions kit - FAQs
Zenon™ Allophycocyanin Mouse IgG2a Labeling Kit, 25 Reactions kit - FAQs
View additional product information for Zenon™ Mouse IgG2a Labeling Kits - FAQs (Z25108, Z25107, Z25155, Z25102, Z25151)
1 product FAQs found
How should I store my allophycocyanin (APC)-conjugated antibody?
Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.