Zenon™ Rabbit IgG Labeling Kits
Invitrogen™

Zenon™ Rabbit IgG Labeling Kits

Generate antibody (IgG) conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ Rabbit IgG Labeling Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityExcitation/EmissionLabel or Dye
Z2531350 Reactions kit402/421 nmAlexa Fluor 405
Z2530250 Reactions kit495/519 nmAlexa Fluor 488
Z25303
also known as Z-25303
50 Reactions kit531/554 nmAlexa Fluor 532
Z2530550 Reactions kit555/565 nmAlexa Fluor 555
Z2530650 Reactions kit578/603 nmAlexa Fluor 568
Z2530750 Reactions kit590/617 nmAlexa Fluor 594
Z2530850 Reactions kit650/668 nmAlexa Fluor 647
Z2535125 Reactions kit650/660 nmAPC (Allophycocyanin)
Z2535525 Reactions kit496, 546, 565/578 nmR-PE (R-Phycoerythrin)
Catalog number Z25313
Price (USD)
409.65
Online exclusive
546.00
Save 136.35
1 kit
Quantity:
50 Reactions kit
Excitation/Emission:
402/421 nm
Label or Dye:
Alexa Fluor 405
Price (USD)
409.65
Online exclusive
546.00
Save 136.35
1 kit
Generate antibody (IgG) conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ Rabbit IgG Labeling Kits. Simplify your laboratory applications and reduce antibody cross-reactivity while also achieving efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Achieve fast, versatile, and reliable fluorophore-, biotin-, or enzyme-labeled IgG primary antibodies with the Zenon™ rabbit IgG labeling kits. These kits utilize Alexa Fluor fluorophores, biotin, Pacific Blue, or enzymes such as R-phycoerythrin and allophycocyanin, which are attached to monovalent, affinity purified Fab fragments. The Fab fragments, in turn, are directed against and bind with the Fc portion of IgG primary antibodies. Only a small amount of starting material is required, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Because the Zenon labeling method is based on immunoselectivity, it does not require the removal of exogenous proteins (such as serum) or amine-containing buffers from the target antibody, simplifying the process.

Zenon labeling technology greatly simplifies the use of multiple mouse-derived antibodies in the same staining protocol. Zenon tricolor labeling kits contain sufficient materials for 10 labeling reactions of each of three different fluorescent colors.

Important features of Zenon labeling technology:
• Labeled antibodies are typically ready to use in 10 minutes
• Requires only 1–20 μg primary antibody
• Simple—no purification required
• Flexible—choose from different fluorophores, biotin, HRP, alkaline phosphatase
• Multiplex with other mouse monoclonal antibodies simultaneously
• Can be used in a variety of applications including ICC, IHC, flow cytometry, and cell imaging.

Advantages of using Zenon antibody labeling kits include:

Cost savings
Zenon antibody labeling kits offer a cost-conscious and reproducible method of tagging as little as 0.4 μg in 2 μL of primary antibody, with minimal waste of expensive or difficult-to-obtain antibodies, or excessive washing steps that pose the risk of product loss.

Sensitivity
Label your primary antibodies without compromising their antigen binding affinity: Zenon dye- and enzyme-labeled Fab fragments, which are targeted to the Fc tail, are affinity purified during their preparation to ensure high affinity and selectivity for the Fc portion of the corresponding primary antibody. The procedure for chemical labeling of the Zenon Fab fragments protects their Fc-binding site, resulting in more active labeling reagents.

Speed
No purification procedure is required prior to using Zenon Fab fragments in your laboratory applications. Formation of the Fab-antibody complex occurs in fewer than five minutes, followed by a five-minute blocking step. During this time, almost all the primary antibody in the mixture is labeled with the labeled Fab fragments.

Simplicity
The Fab-antibody complexes display fluorescence or enzymatic activity that is similar in intensity to that of directly labeled primary antibodies. Varying the extent of the antibody labeling is as simple as changing the amount of added Zenon labeling reagent during the reaction. Once the labeling complexes are formed, they can used immediately, without need for antibody purification.

Reliability
The Zenon Fab-antibody complex is stable and allows subsequent or simultaneous labeling of different target cells and tissues with different complexes. After staining, an aldehyde-based fixing step may be used to prevent the transfer of different labels between different primary antibodies, preserving the initial staining pattern.

Customization
We offer custom antibody conjugation services that are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorViolet
Excitation/Emission402/421 nm
Label TypeAlexa Fluor
Labeling Scale< 1–20 μg
Product LineZenon
Product TypeLabeling Kit
Quantity50 Reactions kit
SpeciesRabbit
Labeling TargetIgG
Label or DyeAlexa Fluor 405
Unit Size1 kit
Contents & Storage
Contains 1 vial of Zenon Alexa Fluor 405 rabbit IgG labeling reagent (250 μL), and 1 vial of Zenon blocking reagent (rabbit IgG, 250 μL).

Store in refrigerator (2–8°C) and protect from light.

Citations & References (2)

Citations & References
Abstract
Optimized multiparametric immunophenotyping of umbilical cord blood cells by flow cytometry.
Authors:Basford C, Forraz N, McGuckin C,
Journal:Nat Protoc
PubMed ID:20595961
Umbilical cord blood is a key source of stem cells for transplantation and regenerative medicine. To maximize each cord blood sample, it is important to analyze its cellular populations. Many cord blood banks focus on counts of total nucleated cells and/or cells that carry the CD34 antigen, but this limited ... More
Glycogen synthase kinase 3beta missplicing contributes to leukemia stem cell generation.
Authors:Abrahamsson AE, Geron I, Gotlib J, Dao KH, Barroga CF, Newton IG, Giles FJ, Durocher J, Creusot RS, Karimi M, Jones C, Zehnder JL, Keating A, Negrin RS, Weissman IL, Jamieson CH,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19237556
Recent evidence suggests that a rare population of self-renewing cancer stem cells (CSC) is responsible for cancer progression and therapeutic resistance. Chronic myeloid leukemia (CML) represents an important paradigm for understanding the genetic and epigenetic events involved in CSC production. CML progresses from a chronic phase (CP) in hematopoietic stem ... More