Fluo-3, AM, cell permeant
Invitrogen™

Fluo-3, AM, cell permeant

Fluo-3, AM, cell permeant

Have Questions?
Catalog NumberQuantity
F142181 mL
Catalog number F14218
Price (USD)
516.00
Each
Quantity:
1 mL
Price (USD)
516.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:

  • Label (Ex/Em of Ca2+–bound form): Fluo-3 (506/526 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: ∼335 nM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Using TPEN to Control Heavy Metal Cations

In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator (Cat. No. TPEN).

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mM
Detection MethodFluorescence
Dye TypeFluo-3
Emission Wavelength526 nm
Excitation Wavelength506 nm
Quantity1 mL
Shipping ConditionRoom Temperature
For Use With (Application)Calcium Indicator
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product TypeDye
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (1252)

Citations & References
Abstract
Calreticulin couples calcium release and calcium influx in integrin-mediated calcium signaling.
Authors:Kwon MS,Park CS,Choi K,Ahnn J,Kim JI,Eom SH,Kaufman SJ,Song WK
Journal:Molecular biology of the cell
PubMed ID:10749940
The engagement of integrin α7 in E63 skeletal muscle cells by laminin or anti-α7 antibodies triggered transient elevations in the intracellular free Ca(2+) concentration that resulted from both inositol triphosphate-evoked Ca(2+) release from intracellular stores and extracellular Ca(2+) influx through voltage-gated, L-type Ca(2+) channels. The extracellular domain of integrin α7 ... More
Cellular alterations produced by the experimental increase in intracellular calcium and the nature of protective effects from pretreatment with nimodipine.
Authors:Danks AM,Hammond DN,Wainer BH,Van Buskirk RG,Isaacson RL
Journal:Brain research. Molecular brain research
PubMed ID:1334195
The immortalized septal cell line, SN56 B5 G4, generated by the fusion of mouse septal area cells and neuroblastoma cells, was used to determine if nimodipine, an antagonist of voltage sensitive calcium 'L' channels, might act in a neuroprotective fashion when intracellular calcium levels were raised by incubation in ouabain ... More
Morphine-3-glucuronide's neuro-excitatory effects are mediated via indirect activation of N-methyl-D-aspartic acid receptors: mechanistic studies in embryonic cultured hippocampal neurones.
Authors:Hemstapat K,Monteith GR,Smith D,Smith MT
Journal:Anesthesia and analgesia
PubMed ID:12873944
Indirect evidence indicates that morphine-3-glucuronide (M3G) may contribute significantly to the neuro-excitatory side effects (myoclonus and allodynia) of large-dose systemic morphine. To gain insight into the mechanism underlying M3G's excitatory behaviors, we used fluo-3 fluorescence digital imaging techniques to assess the acute effects of M3G (5-500 microM) on the cytosolic ... More
Developmental regulation of neuroligand-induced responses in cultured oligodendroglia.
Authors:Belachew S, Malgrange B, Rigo JM, Rogister B, Coucke P, Mazy-Servais C, Moonen G
Journal:Neuroreport
PubMed ID:9601652
Using whole-cell patch-clamp techniques, we show that oligosphere-derived oligodendrocyte progenitor cells (OP) display GABA-, glutamate-, 5-HT-, glycine- and acetylcholine-gated inward currents. When OP differentiate into oligodendrocytes (ODC), the amplitude of peak currents elicited by saturating concentrations of these transmitters decreases except for 5-HT. Intracellular Ca2+ concentration changes induced by microperfusion ... More
Direct detection of uncaged glutamate and the laser photostimulation of cultured rat cortex.
Authors:Torimitsu K, Niwa O
Journal:Neuroreport
PubMed ID:9559923
The photostimulation of nerve cells using a caged compound is very useful because it is non-invasive and non-destructive compared with standard electrophysiological techniques. There are no methods, however, for continuously measuring the photo-uncaged 'free' compound concentration at high temporal and spatial resolutions which can detect how much uncaged compound has ... More